Methyl 3-(chlorosulphonyl)-4-phenyl-5-(trifluoromethyl)thiophene-2-carboxylate
Catalog # APOSPC7614-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:384.778 g/mol
Melting Pt:68…70 °C
Storage Temperature:Ambient
CAS Number:306935-98-8
Formula:C₁₃H₈ClF₃O₄S₂
MDL Number:MFCD02090166