
(1S)-1-[4-(Trifluoromethoxy)phenyl]ethylamine 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:951247-75-9
:
Formula:C₉H₁₀F₃NO
MDL Number:MFCD08057413
:205.177 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together