5-(Hydroxymethyl)-2-methyl-4-(trifluoromethyl)-1,3-thiazole
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1000339-72-9
Formula:C₆H₆F₃NOS
MDL Number:MFCD09264553
:197.178 g/mol
Melting Pt:83…85 °C
Storage Temperature:Ambient
Frequently Bought Together