3'-(Trifluoromethyl)acetophenone oxime
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:99705-50-7
:
Formula:C₉H₈F₃NO
MDL Number:MFCD00085177
:203.161 g/mol
Melting Pt:83…86 °C
Storage Temperature:Ambient
Frequently Bought Together