Order Entry
Czech Republic
ContactUsLinkComponent
1,1'-Carbonylbis(3-methyl-1H-imidazol-3-ium) bis(trifluoromethanesulphonate)
Irritant
1,1'-Carbonylbis(3-methyl-1H-imidazol-3-ium) bis(trifluoromethanesulphonate)
Catalog # APOSPC7301-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1,1'-Carbonylbis(3-methyl-1H-imidazol-3-ium) bis(trifluoromethanesulphonate)
Catalog # APOSPC7301-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :490.356 g/mol
Melting Pt:78…80 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:120418-31-7
Formula:C₉H₁₂N₄O·2CF₃O₃S
MDL Number:MFCD00269944