Methyl 1,6-dihydro-1-(4-fluorophenyl)-6-oxopyridine-3-carboxylate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:929000-81-7
:
Formula:C₁₃H₁₀FNO₃
MDL Number:MFCD09258783
:247.222 g/mol
Storage Temperature:Ambient
Frequently Bought Together