Order Entry
Czech Republic
ContactUsLinkComponent
Methyl 4-fluoro-2-methyl-5-nitrobenzoate 98%
Irritant
Methyl 4-fluoro-2-methyl-5-nitrobenzoate 98%
Catalog # APOSPC6924-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Methyl 4-fluoro-2-methyl-5-nitrobenzoate 98%
Catalog # APOSPC6924-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :213.163 g/mol
Melting Pt:68…71 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:1163287-01-1
Formula:C₉H₈FNO₄
MDL Number:MFCD14581697