
Novec HFE-72DE Engineered Fluid (3M)
Catalog # APOSPC6473-5KG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 kg
- Seq:4114
Specifications
About this item
:
Boiling Pt:45 °C
Density:1.28 g/cm³ (25 °C)
:GHS07
Storage Temperature:Ambient
CAS Number:839682-75-6