4-Methoxy-2-(trifluoromethoxy)benzoyl chloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:886502-42-7
:
Formula:C₉H₆ClF₃O₃
MDL Number:MFCD06660286
:254.59 g/mol
Storage Temperature:Ambient
Frequently Bought Together