
4-Chloro-2-methyl-6-(trifluoromethyl)-1H-benzimidazole
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1211010-30-8
:
Formula:C₉H₆ClF₃N₂
MDL Number:MFCD09027451
:234.606 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together