Order Entry
Czech Republic
ContactUsLinkComponent
2-Methyl-4-(trifluoromethyl)-1,3-thiazole-5-carboxaldehyde 97%
Irritant
2-Methyl-4-(trifluoromethyl)-1,3-thiazole-5-carboxaldehyde 97%
Catalog # APOSPC6115-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-Methyl-4-(trifluoromethyl)-1,3-thiazole-5-carboxaldehyde 97%
Catalog # APOSPC6115-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :195.162 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:1034566-13-6
Formula:C₆H₄F₃NOS
MDL Number:MFCD12198128