

3-(Trifluoromethyl)phenylacetylene 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:705-28-2
:
Formula:C₉H₅F₃
MDL Number:MFCD00467355
:170.131 g/mol
Boiling Pt:146 °C
Density:1.178 g/cm³ (25 °C)
Flash Pt:21 °C
Storage Temperature:Refrigerator
UN:1993
ADR:3,III
:GHS02
:GHS07
Frequently Bought Together