Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Phenyl-4-(trifluoromethyl)-1H-imidazole
2-Phenyl-4-(trifluoromethyl)-1H-imidazole 97%
2-Phenyl-4-(trifluoromethyl)-1H-imidazole 97%
2-Phenyl-4-(trifluoromethyl)-1H-imidazole 97%
APOSPC5741-250MG
:
Apollo Scientific
:
2-Phenyl-4-(trifluoromethyl)-1H-imidazole 97%
APOSPC5741-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
212.171 g/mol
:
Ambient
:
33469-36-2
:
C₁₀H₇F₃N₂
:
MFCD10000861