Order Entry
Czech Republic
ContactUsLinkComponent
1-Bromo-1,1-dichloro-2,2,2-trifluoroethane 96%
1-Bromo-1,1-dichloro-2,2,2-trifluoroethane 96%
Catalog # APOSPC5733-1G
Supplier:  Apollo Scientific
CAS Number:  
1-Bromo-1,1-dichloro-2,2,2-trifluoroethane 96%
Catalog # APOSPC5733-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :231.827 g/mol
Boiling Pt:69…69.2 °C
Storage Temperature:Ambient
CAS Number:354-50-7
Formula:C₂BrCl₂F₃
MDL Number:MFCD01746275