(1R)-N-Methyl-1-[4-(trifluoromethyl)phenyl]ethylamine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:672906-72-8
:
Formula:C₁₀H₁₂F₃N
MDL Number:MFCD07779107
:203.204 g/mol
Storage Temperature:Ambient
Frequently Bought Together