
2,6-Difluoro-3-nitroaniline
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:25892-09-5
:
Formula:C₆H₄F₂N₂O₂
MDL Number:MFCD11110684
:174.105 g/mol
Storage Temperature:Refrigerator
UN:2811
ADR:6.1,III
:GHS07
Frequently Bought Together