
2-Bromo-6-chloro-4-(trifluoromethyl)benzonitrile 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1415130-42-5
:
Formula:C₈H₂BrClF₃N
MDL Number:MFCD27959253
:284.46 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together