Order Entry
Czech Republic
ContactUsLinkComponent
Ethyl 4,4,4-trifluoro-3-hydroxy-2-methylene-3-(trifluoromethyl)butanoate 95%
Irritant
Ethyl 4,4,4-trifluoro-3-hydroxy-2-methylene-3-(trifluoromethyl)butanoate 95%
Catalog # APOSPC56222-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Ethyl 4,4,4-trifluoro-3-hydroxy-2-methylene-3-(trifluoromethyl)butanoate 95%
Catalog # APOSPC56222-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :266.138 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:152251-75-7
Formula:C₈H₈F₆O₃
MDL Number:MFCD28137304