Order Entry
Czech Republic
ContactUsLinkComponent
2-Iodo-4,4,5,5,5-pentafluoropent-1-ol 94%
Irritant
2-Iodo-4,4,5,5,5-pentafluoropent-1-ol 94%
Catalog # APOSPC56213-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-Iodo-4,4,5,5,5-pentafluoropent-1-ol 94%
Catalog # APOSPC56213-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :303.997 g/mol
Boiling Pt:81 °C (13.5 mm Hg)
Density:2.036 g/cm³ (25 °C)
 :GHS07
Storage Temperature:Refrigerator
CAS Number:757-06-2
Formula:C₅H₆F₅IO
MDL Number:MFCD18641885