

2-Chloro-4-iodo-1-(trifluoromethyl)benzene
Catalog # APOSPC56186-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:306.451 g/mol
Storage Temperature:Refrigerator
UN:3265
ADR:8,III
:GHS05
:GHS07
CAS Number:1206599-46-3
Formula:C₇H₃ClF₃I
MDL Number:MFCD12827841