Order Entry
Czech Republic
ContactUsLinkComponent
(R)-1-(2, 4-Difluorophenyl)ethylamine
(R)-1-(2, 4-Difluorophenyl)ethylamine
Catalog # APOSPC56178-1G
Supplier:  Apollo Scientific
CAS Number:  
(R)-1-(2, 4-Difluorophenyl)ethylamine
Catalog # APOSPC56178-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :157.161 g/mol
Storage Temperature:Ambient
CAS Number:791098-84-5
Formula:C₈H₉F₂N
MDL Number:MFCD06761833