
1-[4-(Trifluoromethyl)pyrimidin-2-yl]piperidine-4-carboxamide 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:465514-29-8
:
Formula:C₁₁H₁₃F₃N₄O
MDL Number:MFCD03237056
:274.242 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together