
2,2-Difluoro-2-pyridin-2-ylacetamide ≥95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1064652-41-0
:
Formula:C₇H₆F₂N₂O
MDL Number:MFCD11858603
:172.132 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together