4-Chloro-2-fluorophenylzinc iodide 0.5M in THF
Catalog # APOSPC5552-50ML
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:50 mL
- Seq:2370
Specifications
About this item
:
:321.853 g/mol
Storage Temperature:Ambient
CAS Number:518989-99-6
Formula:C₆H₃ClFIZn
MDL Number:MFCD02260161