
Novec HFE-71DA Engineered Fluid (3M)
Supplier: Apollo Scientific
Certificates
About this item
:
MDL Number:MFCD04040972
Boiling Pt:40 °C
Melting Pt:–29 °C
Density:1.33 g/cm³ (25 °C)
:GHS07
Storage Temperature:Ambient
Frequently Bought Together