Order Entry
Czech Republic
ContactUsLinkComponent
(1S)-1-Cyclopropyl-2,2,2-trifluoroethan-1-amine hydrochloride ≥98%
(1S)-1-Cyclopropyl-2,2,2-trifluoroethan-1-amine hydrochloride ≥98%
Catalog # APOSPC540077-100MG
Supplier:  Apollo Scientific
CAS Number:  
(1S)-1-Cyclopropyl-2,2,2-trifluoroethan-1-amine hydrochloride ≥98%
Catalog # APOSPC540077-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :175.58 g/mol
Storage Temperature:Ambient
CAS Number:1338377-73-3
Formula:C₅H₈F₃N·ClH
MDL Number:MFCD26569801