Order Entry
Czech Republic
ContactUsLinkComponent
6-Chloro-3-(trifluoromethyl)-1H-pyrazolo[4,3-c]pyridine ≥98%
6-Chloro-3-(trifluoromethyl)-1H-pyrazolo[4,3-c]pyridine ≥98%
Catalog # APOSPC540056-100MG
Supplier:  Apollo Scientific
CAS Number:  
6-Chloro-3-(trifluoromethyl)-1H-pyrazolo[4,3-c]pyridine ≥98%
Catalog # APOSPC540056-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :221.567 g/mol
Storage Temperature:Ambient
CAS Number:1431720-68-1
Formula:C₇H₃ClF₃N₃
MDL Number:MFCD28677466