Order Entry
Czech Republic
ContactUsLinkComponent
Methyl 2-amino-3-chloro-5-fluorobenzoate ≥98%
Methyl 2-amino-3-chloro-5-fluorobenzoate ≥98%
Catalog # APOSPC540053-100MG
Supplier:  Apollo Scientific
CAS Number:  
Methyl 2-amino-3-chloro-5-fluorobenzoate ≥98%
Catalog # APOSPC540053-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :203.598 g/mol
Storage Temperature:Ambient
CAS Number:1184351-57-2
Formula:C₈H₇ClFNO₂
MDL Number:MFCD12159877