Order Entry
Czech Republic
ContactUsLinkComponent
(4-(Aminomethyl)-3-fluorophenyl)boronic acid ≥97%
(4-(Aminomethyl)-3-fluorophenyl)boronic acid ≥97%
Catalog # APOSPC540044-250MG
Supplier:  Apollo Scientific
CAS Number:  
(4-(Aminomethyl)-3-fluorophenyl)boronic acid ≥97%
Catalog # APOSPC540044-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :168.961 g/mol
Storage Temperature:Refrigerator
CAS Number:1073055-69-2
Formula:C₇H₉BFNO₂
MDL Number:MFCD24444629