
2,3-Dibromo-6-(trifluoromethyl)pyridine 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1159512-35-2
:
Formula:C₆H₂Br₂F₃N
MDL Number:MFCD11226614
:304.89 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together