
1-(3-Bromo-4-fluoro-phenyl)imidazolidin-2-one
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1566125-49-2
:
Formula:C₉H₈BrFN₂O
MDL Number:MFCD32200331
:259.075 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together