Order Entry
Czech Republic
ContactUsLinkComponent
8-Chloro-6-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridin-3(2H)-one
Irritant
8-Chloro-6-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridin-3(2H)-one
Catalog # APOSPC53585-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
8-Chloro-6-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridin-3(2H)-one
Catalog # APOSPC53585-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :237.566 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1428532-93-7
Formula:C₇H₃ClF₃N₃O
MDL Number:MFCD12406875