
2,2,2-Trifluoro-N-(3-methoxyphenyl)acetamide
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:14818-55-4
:
Formula:C₉H₈F₃NO₂
MDL Number:MFCD00751556
:219.161 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together