
3-Fluoro-4-(methylcarbamoylmethoxy)benzenesulphonyl chloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1557249-94-1
:
Formula:C₉H₉ClFNO₄S
MDL Number:MFCD24328322
ADR:8,III
:281.688 g/mol
UN:1759
:GHS05
Storage Temperature:Ambient
Frequently Bought Together