
1-(2-Bromo-1-methylethoxy)-4-fluorobenzene, tech
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1225544-80-8
:
Formula:C₉H₁₀BrFO
MDL Number:MFCD16316875
:233.078 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together