
3-Methoxy-6-(trifluoromethyl)pyridin-2-amine 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1228898-19-8
:
Formula:C₇H₇F₃N₂O
MDL Number:MFCD16606206
:192.138 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together