
4-Chloro-3-fluoro-2-methylpyridine 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1195251-01-4
:
Formula:C₆H₅ClFN
MDL Number:MFCD16610395
:145.562 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together