Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Expresní objednávka
Stav objednávky
Váš účet
Košík
Czech Republic
Expresní objednávka
Stav objednávky
Váš účet
Košík
ContactUsLinkComponent
Home
3-(Difluoromethyl)-2-fluoroaniline
3-(Difluoromethyl)-2-fluoroaniline 95%
3-(Difluoromethyl)-2-fluoroaniline 95%
APOSPC535020-250MG
:
Apollo Scientific
:
3-(Difluoromethyl)-2-fluoroaniline 95%
APOSPC535020-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
161.124 g/mol
:
GHS07
:
Refrigerator
:
1784796-42-4
:
C₇H₆F₃N
:
MFCD28516613