
1-(4-Chloro-2-fluoro-6-hydroxyphenyl)ethanone 95+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1110663-22-3
:
Formula:C₈H₆ClFO₂
MDL Number:MFCD13152285
:188.583 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together