
2-Fluoro-5-methyl-4-nitrobenzenesulphonyl chloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1803585-04-7
:
Formula:C₇H₅ClFNO₄S
MDL Number:MFCD28797548
ADR:8,III
:253.635 g/mol
UN:3261
:GHS05
Storage Temperature:Ambient
Frequently Bought Together