
Trimethylsilyl 2-chloro-2,2-difluoroacetate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1700654-90-5
:
Formula:C₅H₉ClF₂O₂Si
MDL Number:MFCD30723300
:202.659 g/mol
Storage Temperature:Refrigerator
UN:1760
ADR:8,III
:GHS05
Frequently Bought Together