
2'-Chloro-4'-(trifluoromethoxy)acetophenone
Catalog # APOSPC53279-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:238.591 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1824647-99-5
Formula:C₉H₆ClF₃O₂
MDL Number:MFCD29924117