

2,2,3,3,4,4,4-Heptafluorobutyl chloroformate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:108156-21-4
:
Formula:C₅H₂ClF₇O₂
MDL Number:MFCD25955259
:262.51 g/mol
Boiling Pt:108…109 °C
UN:3277
ADR:6.1+8,II
:GHS05
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together