(S)-1-(5-Chloro-3-fluoropyridin-2-yl)ethanamine
Catalog # APOSPC53144-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:174.603 g/mol
Storage Temperature:Ambient
CAS Number:1932020-62-6
Formula:C₇H₈ClFN₂
MDL Number:MFCD29042492