Order Entry
Czech Republic
ContactUsLinkComponent
 
2-Bromo-4-fluoro-5-nitrotoluene 98%
Catalog # APOSPC53113-5G
Supplier:  Apollo Scientific
CAS Number:  
2-Bromo-4-fluoro-5-nitrotoluene 98%
Catalog # APOSPC53113-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :234.023 g/mol
Storage Temperature:Ambient
CAS Number:64695-96-1
Formula:C₇H₅BrFNO₂
MDL Number:MFCD11847636