2-(Difluoromethyl)imidazo[1,2-a]pyridine-3-carboxylic acid 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2059954-47-9
:
Formula:C₉H₆F₂N₂O₂
MDL Number:MFCD30498045
:212.153 g/mol
Storage Temperature:Ambient
Frequently Bought Together