5-Trifluoromethyl-2H-pyridazin-3-one 95%
Catalog # APOSPC520894-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:164.085 g/mol
Storage Temperature:Ambient
CAS Number:244268-34-6
Formula:C₅H₃F₃N₂O
MDL Number:MFCD09841380