Order Entry
Czech Republic
ContactUsLinkComponent
2,3-Difluoro-1-methyl-4-(trifluoromethoxy)benzene 95%
2,3-Difluoro-1-methyl-4-(trifluoromethoxy)benzene 95%
Catalog # APOSPC520773-1G
Supplier:  Apollo Scientific
CAS Number:  
2,3-Difluoro-1-methyl-4-(trifluoromethoxy)benzene 95%
Catalog # APOSPC520773-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :212.117 g/mol
Storage Temperature:Ambient
CAS Number:1404194-74-6
Formula:C₈H₅F₅O
MDL Number:MFCD22580955