1-(2-Bromo-1,1-difluoroethyl)-2-methoxybenzene 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1186194-40-0
:
Formula:C₉H₉BrF₂O
MDL Number:MFCD12547047
:251.068 g/mol
Storage Temperature:Ambient
Frequently Bought Together