4-(1,1-Difluoroethyl)-1-fluoro-2-(trifluoromethyl)benzene 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1138445-20-1
:
Formula:C₉H₆F₆
MDL Number:MFCD11226712
:228.134 g/mol
Storage Temperature:Ambient
Frequently Bought Together